Skip to Content
Merck
All Photos(1)

Key Documents

WHA10531018

Whatman® protein saver cards

903 Protein saver card (EU), pkg of 100 ea

Synonym(s):

Whatman fta, Z699519, dbs cards, pcr blood

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41111700
NACRES:
NB.79

packaging

pkg of 100 ea

manufacturer/tradename

Whatman 10531018
Whatman Article No. 28416563 (US reference)

Looking for similar products? Visit Product Comparison Guide

General description

Whatman 903 Proteinsaver card is a sample collection card with five half-inch sample spots. Ready-to-use cards and kits save time and costs during protein collection, transport, and archiving.
  • Storage: each circle holds 75-80 µL of sample
  • Strict quality control and ISO9001 manufacturing standards: ensures high quality, reproducibility, and purity

Features and Benefits

  • The Proteinsaver card has a wraparound cover with spaces for name and date of collection.
  • The card fits into our Whatman foil barrier resealable bags for storage.
  • Imprinted with the universal biohazard symbol in accordance with USP regulations.

Other Notes

Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.

Legal Information

Whatman is a registered trademark of Cytiva

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service