Skip to Content
Merck
All Photos(3)

Key Documents

WHAWB120205

QIAcard Non-Indicating FTA Cards

classic, 4 sample areas per card, pkg of 100 cards

Synonym(s):

QIAcard FTA Classic (100), Whatman FTA, Z719730, whatman fta cards

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105500
NACRES:
NB.22

description

FTA classic card with 4 sample areas per card, 125 μL maximum volume/sample area, 500 μL maximum total volume/card

manufacturer/tradename

Qiagen WB120205

packaging

pkg of 100 cards

suitability

suitable for (isolation, purification and storage of nucleic acids)

Looking for similar products? Visit Product Comparison Guide

General description

FTA Cards offer a compact room-temperature storage system that reduces the need for precious freezer space.

QIAcard FTA classic format with 4 spot area to collect, stabilize, process, transport, and archive colored samples such as blood. FTA technology enables cell lysis on contact, denatures proteins and immediate stabilization and protection of nucleic acids.

FTA classic cards utilize QIAcard FTA technology that simplifies the handling and processing of nucleic acids. FTA Cards contain chemicals that lyse cells, denature proteins and protect nucleic acids from nucleases, oxidation and UV damage. FTA cards rapidly inactivate organisms, including blood-borne pathogens, and prevent the growth of bacteria and other microorganisms.
Convenient for protocols that require different locations for testing and archiving samples. Different samples can be processed independently.

Application

Use FTA card technology for a wide range of applications:
  • Transgenic identification
  • Transfusion medicine
  • Plasmid screening
  • Food and agriculture testing
  • Drug discovery
  • Genomics
  • STR analysis
  • Animal identification
  • Whole genome amplification
  • Molecular biology

Features and Benefits

  • 4 sample areas for application of up to 500 μl sample volume per card
  • Simply apply your sample to the FTA Card. Cell membranes and organelles are lysed and the released nucleic acids are entrapped in the fibers of the matrix. The nucleic acids remain immobilized and are preserved for transport, immediate processing or long-term room temperature storage.
  • Since captured nucleic acids are preserved, FTA Cards facilitate sample collection in remote locations and simplify sample transport.
  • Ship your samples back to the laboratory without expensive special handling or dry ice and process at your convenience.
  • FTA Cards can be used with virtually any sample type: Blood, Cultured cells, Buccal cells, Plasmids, and Solid tissue.
  • Captured nucleic acids are ready for purification when you are. Just take a punch from the FTA Card, wash with FTA Purification Reagent and rinse with TE-1 (10 mM Tris-HCI, 0.1 mM EDTA, pH 8) buffer. DNA on the washed punch is ready to use in applications such as PCR, SNP analysis, and real-time PCR. Since PCR products remain in solution, the punch can be used for multiple amplifications.

Other Notes

Sample integrity is optimized when FTA Cards are stored in a Multi-Barrier Pouch with a Desiccant Packet.
Field of Use : For internal research use only

Legal Information

FTA is a trademark of Qiagen Group
QIAcard is a trademark of Qiagen Group

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service