Skip to Content
Merck
All Photos(1)

Key Documents

W290267

Sigma-Aldrich

(-)-α-Pinene

natural, ≥97%, FCC, FG

Synonym(s):

(1S,5S)-2,6,6-Trimethylbicyclo[3.1.1]hept-2-ene, (1S,5S)-2-Pinene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16
CAS Number:
Molecular Weight:
136.23
FEMA Number:
2902
MDL number:
UNSPSC Code:
12164502
Flavis number:
01.004
NACRES:
NA.21
grade:
FG
Fragrance grade
Halal
Kosher
natural
biological source:
Pinus spp.
food allergen:
no known allergens

biological source

Pinus spp.

Quality Level

grade

FG
Fragrance grade
Halal
Kosher
natural

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 872/2012
FCC
FDA 21 CFR 117
FDA 21 CFR 172.515

Assay

≥97%

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

α-pinene and β-pinene

Organoleptic

fresh; camphoraceous; woody; pine

SMILES string

[C@@H]21C([C@@H](C2)C(=CC1)C)(C)C

InChI

1S/C10H16/c1-7-4-5-8-6-9(7)10(8,2)3/h4,8-9H,5-6H2,1-3H3/t8-,9-/m0/s1

InChI key

GRWFGVWFFZKLTI-IUCAKERBSA-N

Related Categories

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

88.3 °F

Flash Point(C)

31.3 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service