Skip to Content
Merck
All Photos(2)

Documents

274399

Sigma-Aldrich

(−)-α-Pinene

99%, optical purity ee: ≥86% (GLC)

Synonym(s):

(-)-alpha-Pinene, (1S)-(−)-α-Pinene, (1S,5S)-2,6,6-Trimethylbicyclo[3.1.1]hept-2-ene, (1S,5S)-2-Pinene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16
CAS Number:
Molecular Weight:
136.23
Beilstein:
1903790
EC Number:
MDL number:
UNSPSC Code:
12352002
eCl@ss:
39011007
PubChem Substance ID:
NACRES:
NA.22

vapor density

4.7 (vs air)

vapor pressure

~3 mmHg ( 20 °C)

Assay

99%

form

liquid

optical activity

[α]20/D −50.0 to −46.0°, neat

optical purity

ee: ≥86% (GLC)

autoignition temp.

491 °F

refractive index

n20/D 1.465 (lit.)

bp

155-156 °C (lit.)

mp

-64 °C (lit.)

density

0.855 g/mL at 25 °C

SMILES string

CC1=CCC2CC1C2(C)C

InChI

1S/C10H16/c1-7-4-5-8-6-9(7)10(8,2)3/h4,8-9H,5-6H2,1-3H3/t8-,9-/m0/s1

InChI key

GRWFGVWFFZKLTI-IUCAKERBSA-N

Looking for similar products? Visit Product Comparison Guide

General description

α-Pinene is a monoterpene used in industries like fragrance, pharmaceuticals, flavor, and cosmetics. It is a major component of pine oil and turpentine oil. Isomerization of α-pinene yields industrially important compounds like camphene, limonene, and p-cymene.

Application

(−)-α-Pinene can be used:
  • To prepare chiral γ-amino alcohols, which are used as catalysts for the preparation of chiral 1-aryl-1-propanols using aromatic aldehydes.
  • As a substrate in the study of its t-BuOOH mediated oxidation catalyzed by Co(II) complex.
  • As a starting material for the synthesis of polyfunctionalized cyclobutane derivatives.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

88.3 °F

Flash Point(C)

31.3 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Isomerization of alpha-pinene over dealuminated ferrierite-type zeolites
Rachwalik R, et al.
J. Catal., 252(2), 161-170 (2007)
Divergent routes to chiral cyclobutane synthons from (−)-alpha-pinene and their use in the stereoselective synthesis of dehydro amino acids
Moglioni AG, et al.
The Journal of Organic Chemistry, 65(13), 3934-3940 (2000)
Co (II) catalysed oxidation of alpha-pinene by molecular oxygen: Part III
Lajunen MK
J. Mol. Catal. A: Chem., 169(1-2), 33-40 (2001)
Enantioselective addition of diethylzinc to aldehydes catalyzed by ?-amino alcohols derived from (+)-and (-)-a-pinene.
Szakonyi Z, et al.
Tetrahedron Asymmetry, 17(2), 199-204 (2006)
Isomerisation of alpha-pinene using modified montmorillonite clays
Yadav MK, et al.
J. Mol. Catal. A: Chem., 216(1), 51-59 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service