Skip to Content
Merck
All Photos(1)

Key Documents

W237418

Sigma-Aldrich

Diethyl malate

≥97%, FG

Synonym(s):

Diethyl 2-hydroxybutanedioate, Hydroxybutane dioic acid diethyl ester, Malic acid diethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5O2CCH2CH(OH)CO2C2H5
CAS Number:
Molecular Weight:
190.19
FEMA Number:
2374
EC Number:
Council of Europe no.:
382
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
09.439
NACRES:
NA.21
Organoleptic:
caramel; fruity; sweet
grade:
FG
Halal
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Halal

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117
FDA 21 CFR 172.515

Assay

≥97%

bp

122-124 °C/12 mmHg (lit.)

density

1.128 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

caramel; fruity; sweet

SMILES string

CCOC(=O)CC(O)C(=O)OCC

InChI

1S/C8H14O5/c1-3-12-7(10)5-6(9)8(11)13-4-2/h6,9H,3-5H2,1-2H3

InChI key

VKNUORWMCINMRB-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

185.0 °F

Flash Point(C)

85 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J M Messana et al.
The American journal of physiology, 255(5 Pt 2), F874-F884 (1988-11-01)
Glutathione, comprising a major portion of cellular nonprotein thiols, plays a central role in a diverse group of cell metabolic functions. Glutathione and related enzyme systems have been shown to protect against both toxin and oxidant-induced injury in several organ
N Chiba et al.
Journal of chromatography. B, Biomedical sciences and applications, 728(1), 35-40 (1999-06-24)
A simple and sensitive method, applicable to quantification of 5-hydroperoxyeicosatetraenoic acid (5-HPETE) produced in cells has been developed using high-performance liquid chromatography on a silica gel column with chemiluminescence detection. 5-HPETE was clearly separated from other positional isomers of HPETEs
Oscar A Bizzozero et al.
Journal of neuroscience research, 83(4), 656-667 (2006-02-01)
This study was aimed at establishing whether oxidative stress induced by acute depletion of brain glutathione (GSH) is sufficient to generate protein carbonyls (PCOs). To this end, rat brain slices were incubated separately with the GSH depletors 1,3-bis[2-chloroethyl]-1-nitrosourea (BCNU) and
H Nishiwaki et al.
Japanese journal of pharmacology, 49(1), 27-35 (1989-01-01)
Effects of adrenalectomy (AD) on ethanol-induced gastric injury and prostaglandin (PG) protection on the damage were investigated in rats and compared with those of N-ethylmaleimide (NEM), a sulfhydryl (SH) blocker, and diethyl maleate (DEM), a SH depletor. Oral administration of
Said H Audi et al.
Journal of applied physiology (Bethesda, Md. : 1985), 113(4), 658-665 (2012-05-26)
Rat exposure to 60% oxygen (O(2)) for 7 days (hyper-60) or to >95% O(2) for 2 days followed by 24 h in room air (hyper-95R) confers susceptibility or tolerance, respectively, of the otherwise lethal effects of subsequent exposure to 100%

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service