Skip to Content
Merck
All Photos(1)

Documents

242926

Sigma-Aldrich

Dimethyl fumarate

97%

Synonym(s):

(2E)-2-Butenedioic acid dimethyl ester, (E)-But-2-enedioic acid dimethyl ester, Allomaleic acid dimethyl ester, Boletic acid dimethyl ester, Dimethyl trans-ethylenedicarboxylate, Methyl fumarate, trans-1,2-Ethylenedicarboxylic acid dimethyl ester, trans-Butenedioic acid dimethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OCOCH=CHCOOCH3
CAS Number:
Molecular Weight:
144.13
Beilstein:
774590
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

bp

192-193 °C (lit.)

mp

102-106 °C (lit.)

SMILES string

[H]\C(=C(\[H])C(=O)OC)C(=O)OC

InChI

1S/C6H8O4/c1-9-5(7)3-4-6(8)10-2/h3-4H,1-2H3/b4-3+

InChI key

LDCRTTXIJACKKU-ONEGZZNKSA-N

Gene Information

human ... KEAP1(9817)

Looking for similar products? Visit Product Comparison Guide

Application

Dimethyl fumarate can be used:
  • In the preparation of an adduct [dimethyl-(+)-(11R,12R)-9,10-dihydro-9,10-ethanoanthracene-11,12-dicarboxylate].
  • As a ligand in the synthesis of Ni(II)-based catalytic system applicable in the Negishi alkylations of N-sulfonyl aziridines with organozinc reagents.
  • As a ligand to synthesize zerovalent ruthenium metal complex namely Ru(η6-1,3,5-cyclooctatriene)(η2-dimethyl fumarate)2.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Aquatic Chronic 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Giovanna Casili et al.
Journal of neurotrauma, 35(13), 1437-1451 (2018-01-25)
Traumatic brain injury (TBI) is a serious neuropathology that causes secondary injury mechanisms, including dynamic interplay between ischemic, inflammatory, and cytotoxic processes. Fumaric acid esters (FAEs) showed beneficial effects in pre-clinical models of neuroinflammation and toxic oxidative stress, so the
Importance of c*--h based modes and large amplitude motion effects in vibrational circular dichroism spectra: the case of the chiral adduct of dimethyl fumarate and anthracene
Passarello M, et al.
The Journal of Physical Chemistry A, 118, 4339-4350 (2014)
Marco Passarello et al.
The journal of physical chemistry. A, 118(24), 4339-4350 (2014-05-21)
The role played by the C*-H based modes (C* being the chiral carbon atom) and the large amplitude motions in the vibrational absorption (VA) and vibrational circular dichroism (VCD) spectra is investigated. The example of an adduct of dimethyl fumarate
Ru (η6-1, 3, 5-cyclooctatriene)(η2-dimethyl fumarate) 2: a novel, versatile zerovalent ruthenium complex with electron-deficient olefinic ligands
Mitsudo T, et al.
Journal of Organometallic Chemistry, 689, 4530-4539 (2004)
Chung-Yang Huang et al.
Journal of the American Chemical Society, 134(23), 9541-9544 (2012-03-15)
A nickel-catalyzed cross-coupling reaction between N-sulfonyl aziridines and organozinc reagents is reported. The catalytic system comprises an inexpensive and air-stable Ni(II) source and dimethyl fumarate as ligand. Regioselective synthesis of β-substituted amines is possible under mild and functional-group-tolerant conditions. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service