Skip to Content
Merck
All Photos(1)

Key Documents

T511609

Sigma-Aldrich

(S)-TCYP

AldrichCPR

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C68H81O4P
Molecular Weight:
993.34
UNSPSC Code:
12161600

Quality Level

reaction suitability

reagent type: ligand

refractive index

n/D

functional group

phosphine

SMILES string

OP1(=O)Oc2c(cc3ccccc3c2-c4c(O1)c(cc5ccccc45)C6CCCCC6)C7CCCCC7

InChI

1S/C32H33O4P/c33-37(34)35-31-27(21-11-3-1-4-12-21)19-23-15-7-9-17-25(23)29(31)30-26-18-10-8-16-24(26)20-28(32(30)36-37)22-13-5-2-6-14-22/h7-10,15-22H,1-6,11-14H2,(H,33,34)

InChI key

RMVWMWZCAKQBAI-UHFFFAOYSA-N

General description

(S)-TCYP is a chiral phosphoric acid catalyst that is commonly used in chemical synthesis. It has been found to be particularly effective in promoting enantioselective reactions. The sterically demanding triscyclohexylbenzene at the 3 and 3′ positions of (S)-TCYP contribute significant steric influence and ressult in its to its high levels of enantioselectivity.

Application

(S)-TCYP has been used in asymmetric aza-ene-type reactions, catalyzing the formation of C-C bonds with high levels of diastereoselectivity

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Understanding the Mechanism of the Asymmetric Propargylation of Aldehydes Promoted by 1,1?-Bi-2-Naphthol-Derived Catalysts
Grayson, M. N.; Goodman, J. M.
Journal of the American Chemical Society, 135, 6142-6148 (2013)
Mechanistic Insights into the BINOL-Derived Phosphoric Acid-Catalyzed Asymmetric Allylboration of Aldehydes
Grayson, M. N.; Pellegrinet, S. C.; Goodman, J. M.
Journal of the American Chemical Society, 134, 2716?2722- 2716?2722 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service