Skip to Content
Merck
All Photos(1)

Key Documents

688320

Sigma-Aldrich

(S)-VAPOL hydrogenphosphate

Synonym(s):

(S)-18-Hydroxy-8,9-diphenyl-diphenanthro[4,3-d:3′,4′-f][1,3,2]dioxaphosphepin-18-oxide, (S)-2,2′-Diphenyl-3,3′-biphenanthryl-4,4′-diyl phosphate, (S)-VAPOL phosphoric acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H25O4P
CAS Number:
Molecular Weight:
600.60
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

form

solid

optical activity

[α]22/D +412°, c = 1 in chloroform

mp

>400 °C

InChI

1S/C40H25O4P/c41-45(42)43-39-35-29(21-19-27-15-7-9-17-31(27)35)23-33(25-11-3-1-4-12-25)37(39)38-34(26-13-5-2-6-14-26)24-30-22-20-28-16-8-10-18-32(28)36(30)40(38)44-45/h1-24H,(H,41,42)

InChI key

YKIJNEDTUDPMNC-UHFFFAOYSA-N

Application

Catalyst for:
  • Benzoyloxylation of (aryl)oxindoles with benzoyl peroxide
  • Enantioselective synthesis of homoaldols and cyclic acetals via kinetic resoln of homoaldols
  • Desymmetrization of mono- and bicyclic N-acyl meso-aziridines by ring opening
  • Highly enantioselective addition of imides to imines to give aminals
  • Friedel-Crafts reaction of pyrroles with benzylidenebenzamides

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service