Skip to Content
Merck
All Photos(1)

Key Documents

695718

Sigma-Aldrich

(R)-3,3′-Bis(9-anthracenyl)-1,1′-binaphthyl-2,2′-diyl hydrogenphosphate

95%

Synonym(s):

(11bR)-2,6-Di-9-anthracenyl-4-hydroxy-dinaphtho[2,1-d:1′,2′-f][1,3,2]dioxaphosphepin-4-oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C48H29O4P
CAS Number:
Molecular Weight:
700.72
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

solid

mp

288-292 °C (D)

storage temp.

−20°C

SMILES string

OP1(=O)Oc2c(cc3CCC=Cc3c2-c4c(O1)c(cc5ccccc45)-c6c7ccccc7cc8ccccc68)-c9c%10ccccc%10cc%11ccccc9%11

InChI

1S/C48H31O4P/c49-53(50)51-47-41(43-35-19-7-1-13-29(35)25-30-14-2-8-20-36(30)43)27-33-17-5-11-23-39(33)45(47)46-40-24-12-6-18-34(40)28-42(48(46)52-53)44-37-21-9-3-15-31(37)26-32-16-4-10-22-38(32)44/h1-5,7-17,19-28H,6,18H2,(H,49,50)

InChI key

SZKDXDJLUYHXLW-UHFFFAOYSA-N

Application

Catalyst used in asymmetric direct alkylation of α-diazoester via C-H bond cleavage.
Catalysts used in enantioselective condensation/amine addition reactions

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Asymmetric Brønsted acid catalysis: catalytic enantioselective synthesis of highly biologically active dihydroquinazolinones.
Magnus Rueping et al.
Angewandte Chemie (International ed. in English), 48(5), 908-910 (2008-12-23)
Daisuke Uraguchi et al.
Journal of the American Chemical Society, 127(26), 9360-9361 (2005-06-30)
A new variant of phosphoric acid-catalyzed C-C bond forming reaction, direct alkylation of alpha-diazoester, via C-H bond cleavage is presented. The resulting products, beta-amino-alpha-diazoesters, are highly functionalized and useful synthetic precursors for various types of beta-amino acids.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service