Skip to Content
Merck
All Photos(3)

Documents

T22209

Sigma-Aldrich

Sulfolane

99%

Synonym(s):

Tetrahydrothiophene 1,1-dioxide, Tetramethylene sulfone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8O2S
CAS Number:
Molecular Weight:
120.17
Beilstein:
107765
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

4.2 (vs air)

Quality Level

vapor pressure

0.01 mmHg ( 20 °C)

Assay

99%

refractive index

n20/D 1.484 (lit.)

bp

104 °C/0.2 mmHg (lit.)
285 °C (lit.)

mp

20-26 °C (lit.)

density

1.261 g/mL at 25 °C (lit.)

SMILES string

O=S1(=O)CCCC1

InChI

1S/C4H8O2S/c5-7(6)3-1-2-4-7/h1-4H2

InChI key

HXJUTPCZVOIRIF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Sulfolane is a polar aprotic solvent that may be used for the complete pyrolysis of cellulose without any polymerization or carbonization side reaction. Sulfolane, in combination with mesitylene, can be used as a solvent in the synthesis of poly(succinimide) (PSI) from L-aspartic acid. It can also be used as a solvent in the synthesis of chlorine-terminated polyethersulfone oligomer, a polymeric activator for nylon 6-polyethersulfone (PES)-nylon 6 block copolymer synthesis.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

350.6 °F - closed cup

Flash Point(C)

177 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thermochemical conversion of cellulose in polar solvent (sulfolane) into levoglucosan and other low molecular-weight substances.
Kawamoto H
Journal of Analytical and Applied Pyrolysis, 70(2), 303-313 (2003)
Christopher Paul Kasanke et al.
Scientific reports, 9(1), 3121-3121 (2019-03-01)
Sulfolane is an industrial solvent and emerging organic contaminant affecting groundwater around the world, but little is known about microbes capable of biodegrading sulfolane or the pathways involved. We combined DNA-based stable isotope probing (SIP) with genome-resolved metagenomics to identify
Polymer micelle-like aggregates of novel amphiphilic biodegradable poly (asparagine) grafted with poly (caprolactone).
Jeong JH
Polymer, 44(3), 583-591 (2003)
Nylon 6-polyethersulfone-nylon 6 block copolymer: synthesis and application as compatibilizer for polyethersulfone/nylon 6 blend.
Ahn TO
Polymer, 38(1), 207-215 (1997)
Self-aggregates of hydrophobically modified poly (2-hydroxyethyl aspartamide) in aqueous solution.
Yang SR
Coll. Polymer Sci., 281(9), 852-861 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service