Skip to Content
Merck
All Photos(4)

Documents

E26258

Sigma-Aldrich

Ethylene carbonate

98%

Synonym(s):

1,3-Dioxolan-2-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H4O3
CAS Number:
Molecular Weight:
88.06
Beilstein:
106249
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

3.04 (vs air)

vapor pressure

0.02 mmHg ( 36.4 °C)

Assay

98%

bp

243-244 °C/740 mmHg (lit.)

mp

35-38 °C (lit.)

density

1.321 g/mL at 25 °C (lit.)

SMILES string

O=C1OCCO1

InChI

1S/C3H4O3/c4-3-5-1-2-6-3/h1-2H2

InChI key

KMTRUDSVKNLOMY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Ethylene carbonate has been used:
  • In the synthesis of aliphatic polyurethanes using diamines and diols.
  • As a precursor for ring-opening polymerization using KOH as initiator.
  • As a reactant in the preparation of dimethyl carbonate, glycerol carbonate by transesterification reaction.

related product

Product No.
Description
Pricing

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - STOT RE 2 Oral

Target Organs

Kidney

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

289.4 °F - closed cup

Flash Point(C)

143 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kinetics of the production of glycerol carbonate by transesterification of glycerol with dimethyl and ethylene carbonate using potassium methoxide, a highly active catalyst.
Esteban J, et al.
Fuel Processing Technology, 138(5), 243-251 (2015)
Ring-opening polymerization of ethylene carbonate and depolymerization of poly (ethylene oxide-co-ethylene carbonate).
Lee J-C and Litt MH
Macromolecules, 33(5), 1618-1627 (2000)
Synthesis of dimethyl carbonate from transesterification of ethylene carbonate with methanol using immobilized ionic liquid on commercial silica.
Kim K-H, et al.
Korean Journal of Chemical Engineering, 27(5), 1441-1445 (2010)
Sang-Jae Park et al.
Journal of the American Chemical Society, 137(7), 2565-2571 (2015-02-04)
Here we describe a class of electric-conducting polymers that conduct electrons via the side chain π-π stacking. These polymers can be designed and synthesized with different chemical moieties to perform different functions, extremely suitable as a conductive polymer binder for
Ahilya N Sawh et al.
Molecular cell, 78(1), 96-111 (2020-02-28)
Current models suggest that chromosome domains segregate into either an active (A) or inactive (B) compartment. B-compartment chromatin is physically separated from the A compartment and compacted by the nuclear lamina. To examine these models in the developmental context of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service