Skip to Content
Merck
All Photos(1)

Key Documents

364525

Sigma-Aldrich

Oleic acid

Oleic acid
1 of 1 reviewers received a sample product or took part in a promotion

technical grade, 90%

Synonym(s):

cis-9-Octadecenoic acid, Elainic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)7CH=CH(CH2)7COOH
CAS Number:
Molecular Weight:
282.46
Beilstein:
1726542
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25
Pricing and availability is not currently available.

biological source

plant

Quality Level

grade

technical grade

vapor pressure

1 mmHg ( 176 °C)

Assay

90%

form

liquid
semisolid
waxy solid

refractive index

n20/D 1.459 (lit.)

bp

194-195 °C/1.2 mmHg (lit.)

mp

13-14 °C (lit.)

solubility

alcohol: soluble
benzene: soluble
chloroform: soluble
diethyl ether: soluble
water: insoluble

density

0.887 g/mL at 25 °C
0.89 g/mL at 25 °C (lit.)

application(s)

microbiology

functional group

oleic acid

lipid type

unsaturated FAs

shipped in

ambient

storage temp.

2-8°C

SMILES string

CCCCCCCC\C=C/CCCCCCCC(O)=O

InChI

1S/C18H34O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h9-10H,2-8,11-17H2,1H3,(H,19,20)/b10-9-

InChI key

ZQPPMHVWECSIRJ-KTKRTIGZSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Oleic acid has been used as a test reagent for studying its impact on the secretion of apoB from HepG2 cells.[1] It has also been used to study the necrophoric response of the southern harvester ant, Pogonomyrmex badius to oleic acid.[2]

Biochem/physiol Actions

Oleic acid is a colourless, odourless fatty acid that blocks the glucose production and food intake when administered intracerebroventricularly.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

D M Gordon
Journal of chemical ecology, 9(1), 105-111 (1983-01-01)
The response of the southern harvester ant,Pogonomyrmex badius, to oleic acid was found to depend on social context. Social context was specified as the number of ants engaging in each of five categories of behavior. When a large percentage of
C R Pullinger et al.
Journal of lipid research, 30(7), 1065-1077 (1989-07-01)
The objective of this study was to establish conditions whereby apoB secreted from HepG2 cells could be regulated over a wide range, and to determine whether changes of output were correlated with the level of apoB mRNA. The presence of
Wasim J Mir et al.
Scientific reports, 7(1), 9647-9647 (2017-08-31)
We investigate the potential use of colloidal nanoplates of Sb
Katarzyna Adamala et al.
Nature protocols, 10(6), 927-938 (2015-05-29)
The liposome dialyzer is a small-volume equilibrium dialysis device, built from commercially available materials, that is designed for the rapid exchange of small volumes of an extraliposomal reagent pool against a liposome preparation. The dialyzer is prepared by modification of

Questions

1–5 of 5 Questions  
  1. If we dilute the stock concentration in PBS, should we continue to store it in 4oC or move it to -20oC? When leaving aliquots of 10 mM stock diluted in 3 mM BSA in PBS, the aliquots precipitated at 4oC, because there's a lower concentration of DMSO.

    1 answer
    1. Unfortunately, there is no stability or storage information available for reconstituted this product.

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  3. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  4. For technical grade, 90% oleic acid, what is the meaning of 90%? What is the remaining 10% made out of?

    1 answer
    1. The minimum purity for Oleic acid content is 90% for this product. The potential impurities in the remaining 10% are not identified. However, other fatty acids such as stearic or linoleic are likely present.

      Helpful?

  5. what is the shelf life of oleic acid (technical grade)?

    1 answer
    1. This product does not contain an expiration date or recommended retest date. Products with no expiration date or recommended retest date should be routinely inspected by customers to ensure they perform as expected. These products are also subject to a one year warranty from the date of shipment. For more information you may access the "Product Dating Information" document under "ADDITIONAL USEFUL DOCUMENTS ABOUT OUR PRODUCTS" at the bottom of the Quality Services page with this link: https://www.sigmaaldrich.com/life-science/quality-and-regulatory-management/quality-services

      Helpful?

Reviews

1 of 1 reviewers received a sample product or took part in a promotion

Active Filters

  1. Stirling, Scotland
    • Review 1
    • Votes 0
    5 out of 5 stars.

    Product ordered

    The quality of the product was good, and the delivery was as scheduled. Really good service which makes my business run more smoothly. In the modern world supply problems are a huge constraint o it was refreshing to get fast and efficient service

    Helpful?

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service