Skip to Content
Merck
All Photos(6)

Documents

KITALYSIS-RCM

Sigma-Aldrich

KitAlysis High-Throughput Medium (5, 6, 7) Ring Closing Metathesis Reaction Screening Kit - Pack of 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161600
NACRES:
NA.22

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling

storage temp.

2-8°C

Application

Screening kit for ring closing metathesis. Step-by-step user guides and a downloadable experimental excel sheets are provided in the below hyperlinks.

Step-by-Step Guide for Ring Closing Metathesis/Cross Metathesis Reaction Screening Kit

Downloadable experimental excel file

Each kit contains 2 customizable screening sets that include the following components:
  • 6x4 pre-weighed catalysts in glass microvials loaded with stir bars and topped with cap mat.
  • 2 (2 mL) ampules each of of ClPh, PhMe, i-PrOAc, Me-THF, and TFA.
  • 1 KitAlysis 24-well reaction block replacement film.
  • 4 empty substrate stock solution reaction vials with stir bars.
  • 1 internal standard.
Monitor reaction progress via TLC-MS by following this protocol

Best when used with KitAlysis Benchtop Inertion Box and 24-well Reaction Block (sold separately).

Features and Benefits

  • Designed and tested by synthetic chemists.
  • Easily run 24-microscale reaction in parallel.
  • 20 μmol reactions with 1 μmol catalyst (5 mol%), 0.2 M concentration, and optional TFA additive.
  • Customizable and downloadable excel template for quick calculations and easy incorporation into electronic laboratory notebooks.

Legal Information

KitAlysis is a trademark of Sigma-Aldrich Co. LLC

Kit Components Only

Product No.
Description

  • SnapIT Ampule Opener, Regular, for 1-2ml, 5-10ml, 10-15ml ampoules 1 EA

Kit Components Also Available Separately

Product No.
Description
SDS

  • 569747Grubbs Catalyst® M204, Umicore 3 µmolSDS

  • 569755Hoveyda-Grubbs Catalyst® M720, Umicore, 97% 3 µmolSDS

  • 579726Grubbs Catalyst® M102, Umicore, 97% 3 µmolSDS

  • 682365Grubbs Catalyst® M207, Umicore 3 µmolSDS

  • 682373Hoveyda-Grubbs Catalyst® M721, Umicore 3 µmolSDS

  • 729345Hoveyda-Grubbs Catalyst® M722, Umicore 3 µmolSDS

  • 900519Isopropyl acetate, anhydrous, ZerO2®, ≥99.6% 8 mLSDS

  • 9005202-Methyltetrahydrofuran, anhydrous, contains 250 ppm BHT as stabilizer, ZerO2®, ≥99.0% 8 mLSDS

  • 900521Chlorobenzene, anhydrous, ZerO2®, 99.8% 8 mLSDS

  • 900522Toluene, anhydrous, ZerO2®, 99.8% 8 mLSDS

  • Z118850Spinbar® magnetic stirring fleas, red, L 10 mm, diam. 3 mm 1 EASDS

See All (11)

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Eye Dam. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Scientist-Led High-Throughput Experimentation (HTE) and Its Utility in Academia and Industry.
Schmink JR, et al.
Aldrichimica Acta, 46, 71-80 (2013)
High-Throughput Experimentation in Catalysis and Materials Science.
Gordillo A, et al.
Ullmann's Encyclopedia of Industrial Chemistry (2014)
Karl D Collins et al.
Nature chemistry, 6(10), 859-871 (2014-09-23)
New organic reactivity has often been discovered by happenstance. Several recent research efforts have attempted to leverage this to discover new reactions. In this Review, we attempt to unify reported approaches to reaction discovery on the basis of the practical
Spencer D Dreher et al.
Journal of the American Chemical Society, 130(29), 9257-9259 (2008-06-28)
Microscale parallel experimentation was used to discover three catalyst systems capable of coupling secondary organotrifluoroborates with sterically and electronically demanding aryl chlorides and bromides. The ensuing results represent the first comprehensive study of alkylboron coupling to aryl chlorides and, in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service