Skip to Content
Merck
All Photos(1)

Key Documents

73022

Sigma-Aldrich

2,6-Diisopropylphenylimido-neophylidene[(S)-(−)-BIPHEN]molybdenum(VI)

ringclosing metathesis catalyst, ≥95.0% (C)

Synonym(s):

(S)-Schrock-Hoveyda Catalyst

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C46H61MoNO2
CAS Number:
Molecular Weight:
755.92
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95.0% (C)

form

solid

storage temp.

2-8°C

SMILES string

CC(C)c1cccc(C(C)C)c1N=[Mo]2(Oc3c(cc(C)c(C)c3-c4c(C)c(C)cc(c4O2)C(C)(C)C)C(C)(C)C)=CC(C)(C)c5ccccc5

InChI

1S/C24H34O2.C12H17N.C10H12.Mo/c1-13-11-17(23(5,6)7)21(25)19(15(13)3)20-16(4)14(2)12-18(22(20)26)24(8,9)10;1-8(2)10-6-5-7-11(9(3)4)12(10)13;1-10(2,3)9-7-5-4-6-8-9;/h11-12,25-26H,1-10H3;5-9H,1-4H3;1,4-8H,2-3H3;/q;;;+2/p-2

InChI key

YLWIQLXPHIAIEN-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Catalyst for the enantioselective olefin metathesis

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G.S. Weatherhead et al.
Tetrahedron Letters, 41, 9553-9553 (2000)
D. S. La et al.
Journal of the American Chemical Society, 120, 9720-9720 (1998)
D S La et al.
Journal of the American Chemical Society, 123(32), 7767-7778 (2001-08-09)
Studies regarding the first examples of catalytic asymmetric ring-opening metathesis (AROM) reactions are detailed. This enantioselective cleavage of norbornyl alkenes is followed by an intermolecular cross metathesis with a terminal olefin partner; judicious selection of olefin is required so that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service