Skip to Content
Merck
All Photos(5)

Key Documents

579726

Sigma-Aldrich

Grubbs Catalyst® M102

Umicore, 97%

Synonym(s):

Benzylidene-bis(tricyclohexylphosphine)dichlororuthenium, Bis(tricyclohexylphosphine)benzylidine ruthenium(IV) dichloride, Dichloro(benzylidene)bis(tricyclohexylphosphine)ruthenium(II), Grubbs Catalyst® 1st Generation, Grubbs Catalyst® M1a (C823)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C43H72Cl2P2Ru
CAS Number:
Molecular Weight:
822.96
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

reaction suitability

core: ruthenium
reagent type: catalyst
reaction type: Ring-Opening Polymerization

mp

153 °C (dec.)

storage temp.

2-8°C

SMILES string

C[Ph].P(C1CCCCC1)(C2CCCCC2)C3CCCCC3.P(C4CCCCC4)(C5CCCCC5)C6CCCCC6.Cl.Cl.[Ru]

Looking for similar products? Visit Product Comparison Guide

Application

First metathesis catalyst to be widely used in organic synthesis. Useful for acyclic diene metathesis polymerization (ADMET), Ring-Opening Metathesis Polymerization (ROMP) of strained cyclic olefins, ring opening metathesis (ROM), olefin cross metathesis (CM) and ring closing metathesis (RCM) of terminal olefins under a variety of reactions conditions.

Learn more about our metathesis catalysts

Legal Information

Product of Umicore

Additional information available at http://www.pmc.umicore.com
Grubbs Catalyst is a registered trademark of Umicore AG & Co. KG

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Sol. 2

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Brummer, O.; Ruckert, a.; Blechert, S.
Chemistry (Weinheim An Der Bergstrasse, Germany), 3, 441-441 (1997)
Michaut, M.; Parrain, J.-L.; Santelli, M.
Chemical Communications (Cambridge, England), 2567-2567 (1998)
Brzezinska, K.; Wolfe, P. S.
Macromolecular Chemistry and Physics, 197, 2065-2065 (1996)
Charvet, R.: Novak, B. M.
Macromolecules, 34, 7680-7680 (2001)
Young-Woo Kim et al.
Nature protocols, 6(6), 761-771 (2011-06-04)
This protocol provides a detailed procedure for the preparation of stapled α-helical peptides, which have proven their potential as useful molecular probes and as next-generation therapeutics. Two crucial features of this protocol are (i) the construction of peptide substrates containing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service