Skip to Content
Merck
All Photos(1)

Key Documents

JKA5101

Sigma-Aldrich

HS-PEG7500-COOH

average MN 7,500, COOH, cross-linking reagent polymerization reactions thiol reactive, cross-linking reagent polymerization reactions amine reactive

Synonym(s):

HS-PEG-COOH, Polyethylene glycol, Thiol-PEG-Carboxyl, HS-PEG-COOH

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HS(CH2CH2O)nCH2COOH
UNSPSC Code:
12162002
NACRES:
NA.23

product name

HS-PEG7500-COOH, average Mn 7,500

form

solid

mol wt

average Mn 7,500

reaction suitability

reagent type: cross-linking reagent
reaction type: Polymerization Reactions
reactivity: amine reactive

reagent type: cross-linking reagent
reaction type: Polymerization Reactions
reactivity: thiol reactive

polymer architecture

shape: linear
functionality: heterobifunctional

General description

Product is a thiolated carboxylic-polyethylene . 4α;-sulfanyl—ω carboxy PEG (HS–PEG–COOH) heterobifunctional PEGs with functional end-groups of very different reactivity. HS-PEG-COOH inhibits nonspecific binding to living cells.

Application

HS-PEG-COOH may be tethered to the gold nanoparticle end during the fabrication of epidermal growth factor (EGF)-conjugated surface-enhanced Raman spectroscopy (SERS) nanoparticles. HS-PEG-COOH was coated onto silicon-on-insulator photonic biosensor chips. Quantum dots (QD)/ virus-like particles (VLPs) could be obtained only from HS-PEG-COOH-coated QDs. It may be used to stabilize nanoparticles and prevent its aggregation.
Applications may include: bioconjugation, drug delivery, PEG hydrogel, crosslinker, and surface functionalization

Legal Information

Product of JenKem Technology

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Antibody-functionalized SERS tags with improved sensitivity.
Guarrotxena N and Bazan GC
Chemical Communications (Cambridge, England), 47(31), 8784-8786 (2011)
SOI optical microring resonator with poly (ethylene glycol) polymer brush for label-free biosensor applications
De Vos, K, et al.
Biosensors And Bioelectronics, 24(8), 2528-2533 (2009)
Quantum dot encapsulation in viral capsids."
Dixit SK, et al.
Nano Letters, 6(9), 1993-1999 (2006)
Min-Xuan Xu et al.
Advanced healthcare materials, 7(17), e1800427-e1800427 (2018-06-27)
The mortality rate of acute liver failure significantly increases due to fatal septicemia. Inactive rhomboid protein 2 (iRhom2) is an essential regulator of shedding TNF-α by trafficking with TNF-α converting enzyme (TACE). Fisetin, a flavonoid present in various fruits and
Tian, L., et al.
Biosensors And Bioelectronics, 59 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service