Skip to Content
Merck
All Photos(1)

Key Documents

QBD10247

Sigma-Aldrich

Thiol-dPEG®4-acid

>95% (HPLC)

Synonym(s):

Carboxy-PEG4-thiol, HS-PEG4-COOH, Thiol-PEG-acid, Thiol-PEG4-COOH, Thiol-PEG4-acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H22O6S
Molecular Weight:
282.35
MDL number:
UNSPSC Code:
12352106
NACRES:
NA.22

Assay

>95% (HPLC)

form

solid or viscous liquid

reaction suitability

reaction type: Pegylations

polymer architecture

shape: linear
functionality: heterobifunctional

shipped in

ambient

storage temp.

−20°C

SMILES string

SCCOCCOCCOCCOCCC(=O)O

InChI

1S/C11H22O6S/c12-11(13)1-2-14-3-4-15-5-6-16-7-8-17-9-10-18/h18H,1-10H2,(H,12,13)

InChI key

ORLGUGSEJFKLED-UHFFFAOYSA-N

Features and Benefits

Thiol-dPEG4-acid is a sulfhydryl-containing, crosslinking PEGylation reagent that has a cross-bridge of monodisperse polyethylene glycol. The single molecular weight, discrete-length PEG (dPEG) chain is 16 atoms (18.3 Å) long. The sulfhydryl end of the crosslinker reacts with gold (forming dative bonds) and with thiol-reactive functional groups such as maleimide, bromoacetyl, SPDP, and thiol. The carboxylic acid end of the molecule couples to free amines using EDC or any suitable carbodiimide. An acylating agent such as N-hydroxysuccinimide (NHS) or 2,3,5,6-tetrafluorophenol (TFP) can enhance coupling efficiency. The carboxylate must react after the sulfhydryl conjugation.

Legal Information

Products Protected under U.S. Patent #s 7,888,536 & 8,637,711 and European Patent #s 1,594,440 & 2,750,681
dPEG is a registered trademark of Quanta BioDesign

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daichi Okuno et al.
Nanoscale, 10(8), 4036-4040 (2018-02-13)
The artificial bilayer single channel recording technique is commonly used to observe the detailed physiological properties of various ion channel proteins. It permits easy control of the solution and membrane lipid composition, and is also compatible with pharmacological screening devices.
Carrie A Simpson et al.
ACS nano, 5(5), 3577-3584 (2011-04-09)
Monolayer-protected gold nanoparticles have great potential as novel building blocks for the design of new drugs and therapeutics based on the easy ability to multifunctionalize them for biological targeting and drug activity. In order to create nanoparticles that are biocompatible
Daichi Okuno et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 32(12), 1353-1357 (2016-12-13)
The artificial bilayer single-channel recording technique is commonly used to observe detailed pharmacological properties of various ion channel proteins. It permits easy control of the solution and membrane lipid composition, and is also compatible with pharmacological screening devices. However, its
Pi-Chou Hsieh et al.
BioMed research international, 2017, 5041683-5041683 (2017-05-02)
Herein, we report a method of combining bioinformatics and biosensing technologies to select aptamers against prostate specific antigen (PSA). The main objective of this study is to select DNA aptamers with higher binding affinity for PSA by using the proposed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service