Skip to Content
Merck
All Photos(3)

Key Documents

757829

Sigma-Aldrich

Poly(ethylene glycol) 2-mercaptoethyl ether acetic acid

average Mn 2,000 (n~45), cross-linking reagent amine reactive, cross-linking reagent thiol reactive

Synonym(s):

Polyethylene glycol, mercaptoPEG acid, mercaptopoly(ethylene glycol) carboxylic acid, mercaptopolyoxyethylene acetic acid, thio-PEG, thiol-PEG-acid, thiol-PEG-carboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HSC2H4O(C2H4O)nCH2CO2H
CAS Number:
UNSPSC Code:
12162002
NACRES:
NA.23

product name

Poly(ethylene glycol) 2-mercaptoethyl ether acetic acid, average Mn 2,100

form

solid

Quality Level

mol wt

PEG average Mn 2,000 (n~45)
average Mn 2,100

reaction suitability

reagent type: cross-linking reagent
reactivity: amine reactive

reagent type: cross-linking reagent
reactivity: thiol reactive

mp

51-56 °C

Ω-end

thiol

α-end

carboxylic acid

polymer architecture

shape: linear
functionality: heterobifunctional

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

Reported applications include: bioconjugation, drug delivery, PEG hydrogel, crosslinker, and surface functionalization.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

>604.4 °F

Flash Point(C)

> 318 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service