Skip to Content
Merck
All Photos(2)

Key Documents

D175005

Sigma-Aldrich

2,6-Dimethylphenol

2,6-Dimethylphenol
1 of 1 reviewers received a sample product or took part in a promotion

99%

Synonym(s):

2-Hydroxy-m-xylene, vic.-m-Xylenol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C6H3OH
CAS Number:
Molecular Weight:
122.16
Beilstein:
1446677
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

99%

form

powder

autoignition temp.

1110 °F

bp

203 °C (lit.)

mp

43-45 °C (lit.)

SMILES string

Cc1cccc(C)c1O

InChI

1S/C8H10O/c1-6-4-3-5-7(2)8(6)9/h3-5,9H,1-2H3

InChI key

NXXYKOUNUYWIHA-UHFFFAOYSA-N

Gene Information

human ... GABRA1(2554)

Looking for similar products? Visit Product Comparison Guide

Related Categories

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

186.8 °F - closed cup

Flash Point(C)

86 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

O Grech-Bélanger et al.
Research communications in chemical pathology and pharmacology, 58(1), 53-62 (1987-10-01)
A new metabolite, 2,6-dimethylphenol obtained by O-dealkylation of mexiletine by rabbit liver was identified. A g.c.-f.i.d. method was developed for its analysis in hepatic homogenates and some of the characteristics of the metabolic reaction were studied. Formation of 2,6-dimethylphenol is
Tingting Li et al.
Talanta, 78(4-5), 1497-1502 (2009-04-14)
In the present work, a simple, selective, and sensitive method has been proposed for the determination of four phenols (catechol, resorcinol, 2,6-dimethylphenol, and 2,4,6-trinitrophenol) in water samples. The method is based on poly-(methacrylic acid-co-ethylene glycol dimethacrylate) monolith microextraction (PMME) and
Gertrud Haeseler et al.
British journal of pharmacology, 145(7), 916-925 (2005-05-25)
Phenol derivatives constitute a family of neuroactive compounds. The aim of our study was to identify structural features that determine their modulatory effects at glycine receptors. We investigated the effects of four methylated phenol derivatives and two halogenated analogues on
L Puig-Grajales et al.
Applied microbiology and biotechnology, 54(5), 692-697 (2000-12-29)
Alkylphenols and fuel oxygenates are important environmental pollutants produced by the petrochemical industry. A batch biodegradability test was conducted with selected ortho-substituted alkylphenols (2-cresol, 2,6-dimethylphenol and 2-ethylphenol), fuel oxygenates (methyl tert-butyl ether, ethyl tert-butyl ether and tert-amylmethyl ether) and tert-butyl
J Hartung et al.
Zentralblatt fur Bakteriologie, Mikrobiologie und Hygiene. 1. Abt. Originale B, Hygiene, 179(5), 431-439 (1984-10-01)
Dust-borne phenols and indols were extracted by ethanol from the sedimentation dust of two pig houses (finishing pigs on half slatted floor resp. piglets on deep litter) and a hen house (3-stage battery cages) and analysed by gas chromatography. In

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

1 of 1 reviewers received a sample product or took part in a promotion

Active Filters

  1. Breda
    • Review 1
    • Votes 0
    5 out of 5 stars.

    Worked flawlessly

    I wouldn't have chosen a different product. Thank you!

    Helpful?

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service