Skip to Content
Merck
All Photos(3)

Key Documents

144134

Sigma-Aldrich

3,5-Dimethylphenol

≥99%

Synonym(s):

5-Hydroxy-m-xylene, sym.-m-Xylenol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C6H3OH
CAS Number:
Molecular Weight:
122.16
Beilstein:
774117
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99%

form

fibers
solid

bp

222 °C (lit.)

mp

61-64 °C (lit.)
65-66 °C

SMILES string

Cc1cc(C)cc(O)c1

InChI

1S/C8H10O/c1-6-3-7(2)5-8(9)4-6/h3-5,9H,1-2H3

InChI key

TUAMRELNJMMDMT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3,5-Dimethylphenol was used in simultaneous determination of urinary concentrations of phenol, o-, p-, m-cresols, 1-, 2-naphthol and xylenol isomers by capillary gas chromatography. It was used as starting reagent for total synthesis of landomycin A, most potent antitumor angucycline antibiotic and optically active polypropionate units.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

179.6 °F

Flash Point(C)

82 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takashi Sugimura et al.
Organic letters, 6(24), 4439-4442 (2004-11-19)
Optically active polypropionate units were synthesized in 9-11 steps from 3,5-dimethylphenol. The sequence consists of the Buchner reaction controlled by a chiral 2,4-pentanediol tether and diastereoselective hydrogenation over Raney nickel. [reaction: see text]
Xiaoyu Yang et al.
Journal of the American Chemical Society, 133(32), 12433-12435 (2011-07-26)
The first total synthesis of landomycin A, the longest and most potent antitumor angucycline antibiotic, has been achieved in 63 steps and 0.34% overall yield starting from 2,5-dihydroxybenzoic acid, 3,5-dimethylphenol, triacetyl d-glucal, and d-xylose, with a convergent linear sequence of
Fang-Jun Huo et al.
The Analyst, 136(9), 1892-1897 (2011-03-05)
A strategy for the determination of the presence of thiol-containing amino acids was successfully established by simply assembling copper chloride and xylenol orange (3,3'-bis[N,N-bis(carboxymethyl)aminomethyl]-o-cresolsulfonephthalein trisodium salt; XO) in a 1 : 1 molar ratio in quasi-physiological water solution (pH 6.0).
A M F Caldeira et al.
Applied radiation and isotopes : including data, instrumentation and methods for use in agriculture, industry and medicine, 65(5), 605-609 (2007-03-14)
Natural oxidation is an undesirable process suffered by gel dosimetry systems as it changes the values and spatial distribution of absorbed doses. Natural oxidation of Fricke xylenol gel (FXG) dosimeter was inferred from optical absorbance values measured for the first
E Södergren et al.
Journal of biochemical and biophysical methods, 37(3), 137-146 (1998-12-31)
The ferrous oxidation in xylenol orange version 2 (FOX2) assay coupled with triphenylphosphine has recently been employed for the measurement of total plasma hydroperoxides (ROOHs). In this study, we have evaluated sample handling and the effect of storage conditions on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service