Recommended Products
Assay
≥95%
form
powder or crystals
reaction suitability
core: nickel
reaction type: Cross Couplings
reagent type: catalyst
mp
>300 °C
SMILES string
O=C(O[Ni]OC(C)=O)C.C1(C2=CC=CC=C2)=CC=NC3=C1C=CC4=C3N=CC=C4C5=CC=CC=C5
Application
Catalyst for cross couplings via C-N bond activation
Caution
Store under N2 atmosphere
related product
Product No.
Description
Pricing
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Journal of the American Chemical Society, 139(15), 5313-5316 (2017-04-01)
We developed a strategy to harness alkyl amines as alkylating agents via C-N bond activation. This Suzuki-Miyaura cross coupling of alkylpyridinium salts, readily formed from primary amines, is the first example of a metal-catalyzed cross coupling via C-N bond activation
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service