Skip to Content
Merck
All Photos(2)

Key Documents

902985

Sigma-Aldrich

(Bathocuproine)NiBr2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H20Br2N2Ni
CAS Number:
Molecular Weight:
578.95
MDL number:
UNSPSC Code:
12161600
NACRES:
NA.22

form

powder or crystals

reaction suitability

core: nickel
reaction type: Cross Couplings
reagent type: catalyst

mp

>300 °C

SMILES string

CC1=NC2=C(C=CC(C(C3=CC=CC=C3)=C4)=C2N=C4C)C(C5=CC=CC=C5)=C1.Br[Ni](C)(C)Br

InChI

1S/C26H20N2.2CH3.2BrH.Ni/c1-17-15-23(19-9-5-3-6-10-19)21-13-14-22-24(20-11-7-4-8-12-20)16-18(2)28-26(22)25(21)27-17;;;;;/h3-16H,1-2H3;2*1H3;2*1H;/q;;;;;+2/p-2

InChI key

DMMFRVQLSPMYOW-UHFFFAOYSA-L

Application

Catalyst for C-alkylation of nitroalkanes with unactivated alkyl iodides

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sina Rezazadeh et al.
Journal of the American Chemical Society, 139(24), 8110-8113 (2017-06-09)
Enabled by nickel catalysis, a mild and general catalytic method for C-alkylation of nitroalkanes with unactivated alkyl iodides is described. Compatible with primary, secondary, and tertiary alkyl iodides; and tolerant of a wide range of functional groups, this method allows

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service