Skip to Content
Merck
All Photos(1)

Key Documents

901079

Sigma-Aldrich

Tin(IV) oxide nanoparticle ink

2.5 wt%, viscosity 3.5 cP 

Synonym(s):

Avantama N-31, Ink for charge selective layers in printed electronics, SnO2 ink, Tin oxide nanoparticle dispersion

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SnO2
UNSPSC Code:
12352303
NACRES:
NA.23
Pricing and availability is not currently available.

Quality Level

description

Solid content: 2.5 wt% crystalline SnO2 in the mixture of butanols

form

dispersion

color

translucent gray-brown

particle size

7-20 nm

viscosity

3.5 cP

Looking for similar products? Visit Product Comparison Guide

Application

  • This SnO2 nanoparticle ink is for charge selective layers in printed electronics.
  • This SnO2 nanoparticle ink is for spin-coating, doctor blading, slot-die or inkjet printing.

Caution

  • Storage: In dark at room temperature.
  • Prior to application: Shake, and filter through 0.2 or 0.45 μm PP filter. Do not use PTFE filter.
  • Ink can be diluted with butanol.
  • Thickness optimization may be necessary.
  • Drying of deposited SnO2 films at >100 °C.

Legal Information

Product of Avantama Ltd.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

84.2 °F - closed cup

Flash Point(C)

29 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service