Skip to Content
Merck
All Photos(4)

Key Documents

549657

Sigma-Aldrich

Tin(IV) oxide

greener alternative

nanopowder, ≤100 nm avg. part. size

Synonym(s):

Tin oxide, Stannic oxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SnO2
CAS Number:
Molecular Weight:
150.71
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

form

nanopowder

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

avg. part. size

≤100 nm

density

6.95 g/mL at 25 °C (lit.)

application(s)

battery manufacturing

greener alternative category

SMILES string

O=[Sn]=O

InChI

1S/2O.Sn

InChI key

XOLBLPGZBRYERU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tin oxide is n type semiconductor with wide band gap. Thermal stability of tin oxide was studied. It′s unique characteristics such as low cost, high gas sensing abilities, low response time and fast recovery makes it a promising material for gas sensors. In addition, it has potential applications in detecting polluted or toxic gases and other species, as well as successful use in optoelectronic devices. Mesoporous tin oxide paste based photo anodes for solar cells. In this process, a printable paste with high viscosity is printed onto semi processed silica wafers using screen printing. This process resulted in integrated microarrays with excellent fabrication yield. Tin oxide nanoparticles may be synthesized by precipitation, hydrothermal, sol gel, hydrolytic, polymeric precursor method and carbothermal reduction.
Tin(IV) oxide nanopowder is a class of electrode material that can be used in the fabrication of lithium-ion batteries. Lithium-ion batteries consist of anode, cathode, and electrolyte with a charge-discharge cycle. These materials enable the formation of greener and sustainable batteries for electrical energy storage.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

A comparative study of nanocrystalline SnO2 materials for thermocatalytic and semiconductor gas sensor applications.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Impact of Molecular Charge-Transfer States on Photocurrent Generation in Solid State Dye-Sensitized Solar Cells Employing Low-Band-Gap Dyes
Raavi SSK, et al.
The Journal of Physical Chemistry C, 118(30), 16825-16830 (2014)
Studies of thermal stability of nanocrystalline SnO2, ZrO2, and SiC for semiconductor and thermocatalytic gas sensors
Pavelko RG, et al.
Russ. J. Electrochem., 45(4), 470-475 (2009)
Water bathing synthesis of high-surface-area nanocrystal-assembled SnO 2 particles.
Masuda Y, et al.
Journal of Solid State Chemistry, 189, 2124-2124 (2012)
Studies of thermal stability of nanocrystalline SnO2, ZrO2, and SiC for semiconductor and thermocatalytic gas sensors
Russ. J. Electrochem., 45(4) (2009)
Comparative study of nanocrystalline SnO 2 materials for gas sensor application: thermal stability and catalytic activity
Sensors and Actuators B, Chemical, 137(2), 637-643 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service