Skip to Content
Merck
All Photos(2)

Documents

700460

Sigma-Aldrich

Indium tin oxide, dispersion

<100 nm particle size (DLS), 30 wt. % in isopropanol

Synonym(s):

ITO

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

form

dispersion

Quality Level

composition

In2O3, 90%
SnO2, 10%

concentration

30 wt. % in isopropanol

particle size

<100 nm (DLS)

bp

82 °C

density

1.04 g/mL at 25 °C

storage temp.

−20°C

SMILES string

O=[Sn]=O.O=[In]O[In]=O

InChI

1S/2In.5O.Sn

InChI key

LNNWKAUHKIHCKO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Indium tin oxide (ITO) is a transparent conductingoxide with good optical transparency (T > 85% at 550 nm) and high electricalconductivity (ρ ∼ 10−4Ωcm). It can be used for various applications that require both opticaltransparency in the visible light region and high electrical conductivity.

Application

Indium tin oxide nanoparticles can be used tofabricate ITO thin films that are applied in the field of electroluminescentdevices, electrochromic systems, liquid crystal display electrodes, solar cells,and energy-efficient windows.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

57.2 °F - closed cup

Flash Point(C)

14 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service