Skip to Content
Merck
All Photos(2)

Key Documents

900680

Sigma-Aldrich

DMT-functionalized silica gel

≥99%, molecular loading ≥0.50 mmol/g

Synonym(s):

2,4,6-Trimercaptotriazine-functionalized silica gel, SiliaMetS® Dimercaptotriazine (DMT)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161600
NACRES:
NA.22

material

silica gel (irregular particle shape)

Quality Level

Assay

≥99%

form

powder

feature

endcapped

reaction suitability

reagent type: catalyst
reagent type: chelator

pore size

60 Å pore diameter

capacity

0.50 mmol/g loading(molecular loading)

bulk density

732 g/L (typical tap density)

compatibility

for use with all common aqueous and organic solvents.

Application

Metal scavenger: Ir, Ni, Os, Pd, Pt, Rh, Ru, Cd, Co, Cu, Fe, Sc, and Zn.

Legal Information

SiliaMetS is a registered trademark of Silicycle, Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service