Skip to Content
Merck
All Photos(1)

Key Documents

SML0791

Sigma-Aldrich

N,N-Dimethyltryptamine

≥97% (HPLC)

Synonym(s):

DMT, Dimethyltryptamine, N,N-Dimethyl-1H-indole-3-ethanamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H16N2
CAS Number:
Molecular Weight:
188.27
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥97% (HPLC)

form

powder

drug control

USDEA Schedule I; regulated under CDSA - not available from Sigma-Aldrich Canada; psicótropo (Spain); Decreto Lei 15/93: Tabela IIA (Portugal)

color

white to beige

solubility

DMSO: 15 mg/mL, clear

storage temp.

−20°C

SMILES string

CN(C)CCC1=CNC2=C1C=CC=C2

InChI

1S/C12H16N2/c1-14(2)8-7-10-9-13-12-6-4-3-5-11(10)12/h3-6,9,13H,7-8H2,1-2H3

InChI key

DMULVCHRPCFFGV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

N,N-Dimethyltryptamine (DMT) functions as a hallucinogen. Experimental studies states that DMT is an endogenous sigma-1 receptor agonist. DMT interacts with sigma-1 receptors and blocks voltage-gated sodium ion (Na+) channels in both native cardiac myocytes and heterologous cells that contain sigma-1 receptors.[1]
N,N-Dimethyltryptamine (DMT) is a hallucinogen now known to be an endogenous sigma-1 receptor agonist. In keeping with the known sigma-1 receptor modulation of voltage gated Na+ channels, DMT was found to reversibly inhibit Na currents by 62% in vitro. N,N-Dimethyltryptamine also induced hypermotility in WT mice (but not in sigma-1 KO mice), all evidence of its sigma-1 agonist activity.
N,N-Dimethyltryptamine is an endogenous sigma-1 receptor agonist; hallucinogen.

Features and Benefits

This compound is a featured product for Neuroscience research. Click here to discover more featured Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Other Notes

Product contains 1.2% wt. EtOH (by NMR and EA).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

STOT SE 3

Target Organs

Central nervous system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ethan H McIlhenny et al.
Biomedical chromatography : BMC, 25(9), 970-984 (2010-11-09)
Ayahuasca, also known as caapi or yage among various South American groups, holds a highly esteemed and millennia-old position in these cultures' medical and religious pharmacopeia. There is now an increasing interest in the potential for modern medical applications of
The Hallucinogen N,N-Dimethyltryptamine (DMT) Is an Endogenous Sigma-1 Receptor Regulator.
Dominique F, et al.
Science, 323(5916), 934-937 (2009)
Rafael G Dos Santos et al.
Psychopharmacology, 219(4), 1039-1053 (2011-08-16)
Ayahuasca is an Amazonian tea containing the natural psychedelic 5-HT(2A/2C/1A) agonist N,N-dimethyltryptamine (DMT). It is used in ceremonial contexts for its visionary properties. The human pharmacology of ayahuasca has been well characterized following its administration in single doses. To evaluate
Tsung-Ping Su et al.
Science signaling, 2(61), pe12-pe12 (2009-03-13)
N,N-dimethyltryptamine (DMT) is a hallucinogen found endogenously in human brain that is commonly recognized to target the 5-hydroxytryptamine 2A receptor or the trace amine-associated receptor to exert its psychedelic effect. DMT has been recently shown to bind sigma-1 receptors, which
Arturo A Vitale et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 52(6), 970-977 (2011-05-31)
N,N-dimethyltryptamine (DMT), a strong psychodysleptic drug, has been found in higher plants, shamanic hallucinogenic beverages, and the urine of schizophrenic patients. The aim of this work was to gain better knowledge on the relationship between this drug and hallucinogenic processes

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service