Skip to Content
Merck
All Photos(3)

Key Documents

538086

Sigma-Aldrich

3-Mercaptopropyl-functionalized silica gel

200-400 mesh, extent of labeling: 1.2 mmol/g loading

Synonym(s):

Mercapto-3 functionalized silica gel

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352309
PubChem Substance ID:
NACRES:
NA.22

reaction suitability

reagent type: catalyst
reagent type: chelator

extent of labeling

1.2 mmol/g loading

surface area

500 m2/g

particle size

200-400 mesh

pore size

60 Å pore size

SMILES string

O=[SiH2].CCCS

InChI key

XVEMJYXZUPRUMO-UHFFFAOYSA-N

Application

3-Mercaptopropyl-functionalized silica gel (MPS) is a resin which can be used:
  • As a binding agent in diffusive gradients in thin film (DGT) technique for the determination of labile Hg, methylmercury and arsenic (AsIII) concentrations.
  • As a heterogeneous support for immobilizing titanocene catalysts for hydrosilylation of 2-phenylpyrroline.
  • As a reducing agent for sulfoxide deoxygenation.
  • As a metal scavenger for various metals such as Pd, Pt, Cu, Ag and Pb.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Speciation of dissolved inorganic arsenic by diffusive gradients in thin films: selective binding of AsIII by 3-mercaptopropyl-functionalized silica gel
Bennett WW, et al.
Analytical Chemistry, 83(21), 8293-8299 (2011)
Measurements of dissolved methylmercury in natural waters using diffusive gradients in thin film (DGT)
Clarisse O and Hintelmann H
Journal of Environmental Monitoring, 8(12), 1242-1247 (2006)
Tethered ansa-bridged titanium complexes immobilized on 3-mercaptopropyl-functionalized silica gel and their application for the hydrosilylation of imines
Gruber-Woelfler H, et al.
Dalton Transactions, 41(41), 12711-12719 (2012)
Strong evidence of solution-phase catalysis associated with palladium leaching from immobilized thiols during Heck and Suzuki coupling of aryl iodides, bromides, and chlorides
Richardson JM and Jones CW
J. Catal., 251(1), 80-93 (2007)
Cathleen M Crudden et al.
Journal of the American Chemical Society, 127(28), 10045-10050 (2005-07-14)
The functionalization of SBA-15 with mercaptopropyl trimethoxysilane leads to a material capable of absorbing Pd from organic and aqueous solutions. The resulting Pd-loaded material acts as a catalyst for the Suzuki-Miyaura and Mizoroki-Heck coupling reactions. Leaching studies show that the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service