Skip to Content
Merck
All Photos(2)

Documents

747793

Sigma-Aldrich

(Ir[dF(CF3)ppy]2(dtbpy))PF6

Synonym(s):

[Ir{dF(CF3)ppy}2(dtbpy)]PF6, [4,4′-Bis(1,1-dimethylethyl)-2,2′-bipyridine-N1,N1′]bis[3,5-difluoro-2-[5-(trifluoromethyl)-2-pyridinyl-N]phenyl-C]Iridium(III) hexafluorophosphate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H34F16IrN4P
CAS Number:
Molecular Weight:
1121.91
UNSPSC Code:
12161600
NACRES:
NA.22

form

solid

Quality Level

reaction suitability

core: iridium
reagent type: catalyst
reaction type: Photocatalysis

mp

>300 °C

photocatalyst activation

450 nm

SMILES string

F[P](F)(F)(F)(F)F.CC(C)(C)C1=CC=[N@H]2C(=C1)C3=CC(=CC=[N@@H]3[Ir]2456c7cc(F)cc(F)c7C8=CC=C(C=[N]48)C(F)(F)F)C(C)(C)C.Fc9cc(F)c(C%10=[N]5C=C(C=C%10)C(F)(F)F)c6c9

Looking for similar products? Visit Product Comparison Guide

Application

(Ir[dF(CF3)ppy]2(dtbpy))PF6 is a cyclometalated iridium(III) complex that can be used in visible-light mediated photocatalytic organic transformations.It has been used to achieve late-stage incorporation of alkyl groups to a variety of biologically active heterocycles.

Product can be used with our line of photoreactors: Including Penn PhD (Z744035) & SynLED 2.0 (Z744080)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Advances in Photocatalysis: A Microreview of Visible Light Mediated Ruthenium and Iridium Catalyzed Organic Transformations.
Teegardin K, et al.
Organic Process Research & Development, 20(7), 1156-1163 (2016)
A photochemical strategy for lignin degradation at room temperature.
Nguyen J D, et al.
Journal of the American Chemical Society, 136(4), 1218-1221 (2014)
Formal Total Synthesis of l-Ossamine via Decarboxylative Functionalization Using Visible-Light-Mediated Photoredox Catalysis in a Flow System.
Inuki S, et al.
The Journal of Organic Chemistry, 82(2), 1248-1253 (2017)
Radical Carbon?Carbon Bond Formations Enabled by Visible Light Active Photocatalysts.
Wallentin C J, et al.
Chimia, 66(6), 394-398 (2012)
Late?Stage Functionalization of Biologically Active Heterocycles Through Photoredox Catalysis.
DiRocco D A et al.
Angewandte Chemie (International Edition in English), 53(19), 4802-4806 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service