804215
[Ir{dFCF3ppy}2(bpy)]PF6
Synonym(s):
[Ir[dF(CF3)2ppy]2(bpy)]PF6
About This Item
Recommended Products
form
powder
Quality Level
reaction suitability
core: iridium
reagent type: catalyst
reaction type: Photocatalysis
photocatalyst activation
450 nm
SMILES string
FP(F)(F)(F)F.FC1=CC(F)=C(C2=[N]3C=C(C(F)(F)F)C=C2)C([Ir+]345([N]6=C(C7=CC=CC=[N]75)C=CC=C6)[N]8=C(C9=C(F)C=C(F)C=C94)C=CC(C(F)(F)F)=C8)=C1.[F-]
InChI
1S/2C12H5F5N.C10H8N2.F5P.FH.Ir/c2*13-8-2-3-9(10(14)5-8)11-4-1-7(6-18-11)12(15,16)17;1-3-7-11-9(5-1)10-6-2-4-8-12-10;1-6(2,3,4)5;;/h2*1-2,4-6H;1-8H;;1H;/q;;;;;+1/p-1
InChI key
DZIQZTFRUCLPRL-UHFFFAOYSA-M
Application
Product can be used with our line of photoreactors: Including Penn PhD (Z744035) & SynLED 2.0 (Z744080)
related product
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service