745952
Barium titanate(IV)
nanopowder (cubic), 50 nm (SEM), 99.9% trace metals basis
Synonym(s):
BaTiO3 nanoparticles, Barium titanium oxide, Barium titanium trioxide, Titanium barium oxide
About This Item
Recommended Products
Assay
99.9% trace metals basis
form
nanopowder (cubic)
particle size
50 nm (SEM)
density
6.08 g/mL at 25 °C (lit.)
SMILES string
[Ba++].[O-][Ti]([O-])=O
InChI
1S/Ba.3O.Ti/q+2;;2*-1;
InChI key
WNKMTAQXMLAYHX-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Inhalation - Acute Tox. 4 Oral
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service