Skip to Content
Merck
All Photos(3)

Key Documents

633844

Sigma-Aldrich

Zinc iron oxide

nanopowder, <100 nm particle size (BET), >99% trace metals basis

Synonym(s):

Diiron zinc tetraoxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ZnFe2O4
CAS Number:
Molecular Weight:
241.08
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

>99% trace metals basis

form

nanopowder

particle size

<100 nm (BET)

SMILES string

O=[Fe]O[Zn]O[Fe]=O

InChI

1S/2Fe.4O.Zn

InChI key

WGEATSXPYVGFCC-UHFFFAOYSA-N

General description

Zinc iron oxide nanoparticles are chemically stable and exhibit superparamagnetic properties. There are a number of methods to prepare ZnO nanoparticles, some of them are self-propagated combustion of fuels (alanine, glycine and proline), co-precipitation, sol-gel reaction, spin coating, ball milling and auto combustion.

Application

Potential applications of ZnFe2O4 are gas sensors, catalysts and MRI contrast agents.

Legal Information

Product of Engi-Mat Co.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Influence of fuel on phase formation of ZnFe 2 O 4 prepared by self-propagated combustion route
Venkatesan K and Babu D
AIP Conference Proceedings, 1665(1) (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service