Skip to Content
Merck
All Photos(2)

Documents

637602

Sigma-Aldrich

Barium ferrite

nanopowder, <100 nm particle size (BET), >97% trace metals basis

Synonym(s):

Barium dodecairon nonadecaoxide, Barium iron oxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BaFe12O19
CAS Number:
Molecular Weight:
1111.46
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Assay

>97% trace metals basis

form

nanopowder

particle size

<100 nm (BET)

SMILES string

O=[Ba].O=[Fe]O[Fe]=O.O=[Fe]O[Fe]=O.O=[Fe]O[Fe]=O.O=[Fe]O[Fe]=O.O=[Fe]O[Fe]=O.O=[Fe]O[Fe]=O

InChI

1S/Ba.12Fe.19O

InChI key

AJCDFVKYMIUXCR-UHFFFAOYSA-N

Legal Information

Product of Engi-Mat Co.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - STOT RE 2 - STOT SE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Barium ferrite nanoparticles prepared directly by aerosol pyrolysis
Gonzalez-Carreno T, et al.
Materials Letters, 43(3), 97-101 (2000)
Structure and magnetic properties of nanoparticles of barium ferrite synthesized using microemulsion processing
Pillai V, et al.
Colloids and Surfaces. A, Physicochemical and Engineering Aspects, 80(1), 69-75 (1993)
A M Granov et al.
Vestnik rentgenologii i radiologii, (5)(5), 53-54 (1993-09-01)
Various types of x-ray endovascular embolization were carried out in 256 patients with renal parenchymatous carcinoma over the period of 1979 to 1991. Iodolipol chemoembolization with dioxadet, a fat-soluble preparation, significantly improved three- and five-year cumulative survival of operable patients
[Specialized outpatient care in infectious diseases].
E N Antakova et al.
Vrachebnoe delo, (7)(7), 113-115 (1982-07-01)
Elif Tümay Özer et al.
Journal of hazardous materials, 229-230, 20-28 (2012-07-04)
The barium hexaferrite (BaFe(12)O(19)) containing magnetic poly(ethylene glycol dimethacrylate-vinyl pyridine), (mag-poly(EGDMA-VP)) beads (average diameter=53-212 μm) were synthesized and characterized. Their use as an adsorbent in the removal of diethyl phthalate (DEP) from an aqueous solution was investigated. The mag-poly(EGDMA-VP) beads

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service