734616
Hexa(ethylene glycol) dithiol
>97%, cross-linking reagent thiol reactive
Synonym(s):
Polyethylene glycol, 3,6,9,12,15-Pentaoxaheptadecane-1,17-dithiol, Tetraethylene glycol di(ethanediol)
About This Item
Recommended Products
product name
Hexa(ethylene glycol) dithiol, >97%
Quality Level
Assay
>97%
form
liquid
reaction suitability
reagent type: cross-linking reagent
reactivity: thiol reactive
refractive index
n20/D 1.495
density
1.113 g/mL at 25 °C
polymer architecture
shape: linear
functionality: homobifunctional
storage temp.
2-8°C
SMILES string
SCCOCCOCCOCCOCCOCCS
InChI
1S/C12H26O5S2/c18-11-9-16-7-5-14-3-1-13-2-4-15-6-8-17-10-12-19/h18-19H,1-12H2
InChI key
DMHSDYYORJTHAS-UHFFFAOYSA-N
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service