Skip to Content
Merck
All Photos(3)

Key Documents

381462

Sigma-Aldrich

Pentaerythritol tetrakis(3-mercaptopropionate)

>95%

Synonym(s):

Pentaerythritol (3-mercaptopropionate), Pentaerythritol terakis(3-mercaptopropionate), Pentaerythritol tetra(3-mercaptopropionate)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HSCH2CH2COOCH2)4C
CAS Number:
Molecular Weight:
488.66
Beilstein:
2312625
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Quality Level

Assay

>95%

refractive index

n20/D 1.531 (lit.)

bp

275 °C/1 mmHg (lit.)

density

1.28 g/mL at 25 °C (lit.)

SMILES string

SCCC(=O)OCC(COC(=O)CCS)(COC(=O)CCS)COC(=O)CCS

InChI

1S/C17H28O8S4/c18-13(1-5-26)22-9-17(10-23-14(19)2-6-27,11-24-15(20)3-7-28)12-25-16(21)4-8-29/h26-29H,1-12H2

InChI key

JOBBTVPTPXRUBP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Pentaerythritol tetrakis(3-mercaptopropionate) can be used as a precursor to synthesize:
  • Polymeric degradable networks through thiol-ene click reactions with tri/tetra-acrylates.
  • Thiol-ene-methacrylate composites, which are applicable as dental restorative materials.
  • Network solid polymer electrolytes based on polydimethylsiloxane, for lithium-ion batteries.
It can also be used to functionalize poly(high internal phase emulsions) for removal of heavy metals from water.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gowtham Sathyanarayanan et al.
Analytical and bioanalytical chemistry, 410(25), 6677-6687 (2018-08-04)
We report the development and characterization of digital microfluidic (DMF) immobilized enzyme reactors (IMERs) for studying cytochrome P450 (CYP)-mediated drug metabolism on droplet scale. The on-chip IMERs consist of porous polymer (thiol-ene) monolith plugs prepared in situ by photopolymerization and
Manju Saraswathy et al.
Journal of the mechanical behavior of biomedical materials, 74, 296-303 (2017-06-27)
Significant efforts have been expended to mitigate plasticizer migration from crosslinked methacrylic and poly(vinyl chloride) polymer networks by synthesizing reactive plasticizers that can blend homogenously within the networks to reduce polymer property change, acute toxicity and downstream environmental effects of
Claudio Russo et al.
Polymers, 11(6) (2019-06-07)
Mechanical and rheological properties of novel dual-curing system based on sequential thiol-acrylate and thiol-epoxy reactions are studied with the aim of addressing the obtained materials to suitable advanced applications. The crosslinking process is studied by rheological analysis in order to
Mei Chen et al.
ACS applied materials & interfaces, 9(27), 23246-23254 (2017-06-15)
A series of sticky superhydrophobicity surfaces with high water contact angle and high water adhesive force is facilely prepared via an all-solution-processed method based on polymerization-induced phase separation between liquid crystals (LCs) and epoxy resin, which produces layers of epoxy
N A Traugutt et al.
Soft matter, 13(39), 7013-7025 (2017-09-21)
This study presents the first direct comparison of the influence of liquid-crystal order during synthesis on the thermo-mechanical behaviors of main-chain liquid-crystal elastomers (LCEs) in thiol-acrylate networks. Six polydomain nematic elastomer (PNE) chemistries were compared directly by synthesizing with the

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service