Skip to Content
Merck
All Photos(1)

Key Documents

309583

Sigma-Aldrich

Pentaethylene glycol di(p-toluenesulfonate)

95%

Synonym(s):

3,6,9,12-Tetraoxatetradecane-1,14-diyl ditosylate, Pentaethylene glycol ditosylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4SO3(CH2CH2O)5SO2C6H4CH3
CAS Number:
Molecular Weight:
546.65
Beilstein:
3599245
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

viscous liquid

refractive index

n20/D 1.524 (lit.)

density

1.26 g/mL at 25 °C (lit.)

SMILES string

Cc1ccc(cc1)S(=O)(=O)OCCOCCOCCOCCOCCOS(=O)(=O)c2ccc(C)cc2

InChI

1S/C24H34O10S2/c1-21-3-7-23(8-4-21)35(25,26)33-19-17-31-15-13-29-11-12-30-14-16-32-18-20-34-36(27,28)24-9-5-22(2)6-10-24/h3-10H,11-20H2,1-2H3

InChI key

BUHGDYPBQWWWQS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Pentaethylene glycol di(p-toluenesulfonate) was used in the preparation of 3′-formylbenzo-18-crown-6 by undergoing coupling reaction with 2,3-dihydroxybenzaldehyde. It was also used to prepare calix[4]arenes, calix[5]arenes and crown ethers.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Tetrahedron, 49, 6019-6019 (1993)
Mubarak Ali et al.
Langmuir : the ACS journal of surfaces and colloids, 33(36), 9170-9177 (2017-08-11)
We demonstrate experimentally and theoretically a nanofluidic device for the selective recognition of the cesium ion by exploiting host-guest interactions inside confined geometry. For this purpose, a host molecule, i.e., the amine-terminated p-tert-butylcalix[4]arene-crown (t-BuC[4]C-NH
Chem Express, 8, 471-471 (1993)
Pure and Applied Chemistry. Chimie Pure Et Appliquee, 65, 585-585 (1993)
Complexation and Transport of Zwitterionic Amino Acids by an Artificial Receptor.
Bull. Korean Chem. Soc., 20, 129-131 (1999)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service