Skip to Content
Merck
All Photos(2)

Key Documents

730882

Sigma-Aldrich

Disodium bis(4-chloro-3-sulfophenyl)sulfone

greener alternative

97%

Synonym(s):

Disodium 4,4′-dichloro-3,3′-sulfodiphenyl sulfone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H6Cl2Na2O8S3
CAS Number:
Molecular Weight:
491.25
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

crystals

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

>300 °C

greener alternative category

SMILES string

[Na+].[Na+].[O-]S(=O)(=O)c1cc(ccc1Cl)S(=O)(=O)c2ccc(Cl)c(c2)S([O-])(=O)=O

InChI

1S/C12H8Cl2O8S3.2Na/c13-9-3-1-7(5-11(9)24(17,18)19)23(15,16)8-2-4-10(14)12(6-8)25(20,21)22;;/h1-6H,(H,17,18,19)(H,20,21,22);;/q;2*+1/p-2

InChI key

KKEBUZUONXHUNE-UHFFFAOYSA-L

General description

Disodium bis(4-chloro-3-sulfophenyl)sulfone (SDCDPS) is a hydrophilic sulfonic monomer, which can be synthesized by electrophilic aromatic substitution by sulfonating the 4,4′-dichlorodiphenyl sulfone (DCDPS) monomer.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

SDCDPS can be used in the formation of a sulfonated membrane for the fabrication of proton exchange membrane fuel cells. It can also be used in the synthesis of a stable solid acid catalyst for esterification.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sulfonated polyethersulfone directly synthesized through sulfonic monomer as a new stable solid acid catalyst for esterification
Wang Z, et al.
Catalysis Communications, 27, 164-168 (2012)
Organic-inorganic composite membrane based on sulfonated poly (arylene ether ketone sulfone) with excellent long-term stability for proton exchange membrane fuel cells
Li J, et al.
Journal of Membrane Science , 529, 243-251 (2017)
Compositional effect on the properties of sulfonated and nonsulfonated polymer blend membranes for direct methanol fuel cell
Kim HK, et al.
Macromolecular Research, 19(9), 928-928 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service