Skip to Content
Merck
All Photos(1)

Key Documents

723169

Sigma-Aldrich

Bis[2-(2′-bromoisobutyryloxy)ethyl]disulfide

Synonym(s):

(BiBOE)2S2, ATRP (Bio)degradable Initiator, BiBOEDS

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H20Br2O4S2
CAS Number:
Molecular Weight:
452.22
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

liquid

refractive index

n20/D 1.529

density

1.478 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

CC(C)(Br)C(=O)OCCSSCCOC(=O)C(C)(C)Br

InChI

1S/C12H20Br2O4S2/c1-11(2,13)9(15)17-5-7-19-20-8-6-18-10(16)12(3,4)14/h5-8H2,1-4H3

InChI key

PXBWWYBXPGZCHC-UHFFFAOYSA-N

Application

Atom Transfer Radical Polymerisation (ATRP) initiator for the preparation of biodegradable polymers as well as polymers that adhere to gold surfaces. May also be used to introduce a temperature and light sensitive cleavable region into the polymer.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tsarevsky, N.V.; Matyjaszewski, K.
Macromolecules, 35, 9009-9009 (2002)
Wenjing Huang et al.
Theranostics, 9(13), 3825-3839 (2019-07-10)
Reversing multidrug resistance (MDR) remains a big challenge in cancer therapy. Combining the hyperthermia and chemotherapy is a promising strategy for efficient cancer treatment with MDR reversal. Gold nanocages (GNCs) are an ideal photothermal (PTT)-chemotherapy integration platform due to their
Hao Zhao et al.
ACS nano, 13(6), 6647-6661 (2019-05-16)
Highly efficient nanoarchitectures are of great interest for achieving precise chemotherapy with minimized adverse side effects in cancer therapy. However, a major challenge remains in exploring a rational approach to synthesize spatiotemporally selective vehicles for precise cancer chemotherapy. Here, we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service