Skip to Content
Merck
All Photos(3)

Documents

719447

Sigma-Aldrich

XtalFluor-M®

Synonym(s):

Difluoro(morpholino)sulfonium tetrafluoroborate, Difluoro-4-morpholinylsulfonium tetrafluoroborate, Morpho-DAST difluorosulfinium salt, Morpholinodifluorosulfinium tetrafluoroborate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8BF6NOS
CAS Number:
Molecular Weight:
242.98
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

form

solid

Quality Level

mp

117-126 °C

storage temp.

−20°C

SMILES string

F[B-](F)(F)F.F\S(F)=[N+]1/CCOCC1

InChI

1S/C4H8F2NOS.BF4/c5-9(6)7-1-3-8-4-2-7;2-1(3,4)5/h1-4H2;/q+1;-1

InChI key

SOCYGFRGCVLCSU-UHFFFAOYSA-N

Application

Convenient crystalline highly chemoselective deoxofluorination reagent with a broad substrate scope

Deoxofluorination reagent with a better safety profile, that doesn′t generate corrosive HF which makes it suitable for use in standard borosilicate vessels, and does not react violently with water

Reactant for:
  • Preparation of fluorodisaccharides

Legal Information

XtalFluor-M is a registered trademark of OmegaChem, Inc.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Francis Beaulieu et al.
Organic letters, 11(21), 5050-5053 (2009-10-06)
Aminodifluorosulfinium tetrafluoroborate salts were found to act as efficient deoxofluorinating reagents when promoted by an exogenous fluoride source and, in most cases, exhibited greater selectivity by providing less elimination byproduct as compared to DAST and Deoxo-Fluor. Aminodifluorosulfinium tetrafluoroborates are easy
Alexandre L'heureux et al.
The Journal of organic chemistry, 75(10), 3401-3411 (2010-04-22)
Diethylaminodifluorosulfinium tetrafluoroborate (XtalFluor-E) and morpholinodifluorosulfinium tetrafluoroborate (XtalFluor-M) are crystalline fluorinating agents that are more easily handled and significantly more stable than Deoxo-Fluor, DAST, and their analogues. These reagents can be prepared in a safer and more cost-efficient manner by avoiding

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service