Skip to Content
Merck
All Photos(1)

Key Documents

802794

Sigma-Aldrich

DMPU-HF Reagent

Synonym(s):

DMPU-HF Complex (HF 65% w/w)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H13FN2O
CAS Number:
Molecular Weight:
148.18
UNSPSC Code:
12352000
PubChem Substance ID:
NACRES:
NA.22

form

liquid

Quality Level

storage temp.

2-8°C

SMILES string

F/O=C1N(C)CCCN\1C

InChI

1S/C6H12FN2O/c1-8-4-3-5-9(2)6(8)10-7/h3-5H2,1-2H3

InChI key

ZTQIVQDBIIEDTI-UHFFFAOYSA-N

Application

DMPU-HF Reagent is a nucleophilic fluorination reagent that can be used:       
  • To prepare fluoroalkenes and gem-difluoromethlylene from substituted alkynes in the presence of an Au-JohnPhos complex as a catalyst.      
  • To synthesize substituted 4-fluorotetrahydropyrans and 4-fluoropiperidines from homoallylic alcohol/amine with aldehyde via fluoro-Prins reaction.       
  • In the bromofluorination of alkenes in the presence of N-bromosuccinimide to yield corresponding bromofluoro compounds.

DMPU-HF, like TREAT-HF and Olah′s Reagent, is a convenient way of utilizing Hydrogen Fluoride as a fluorination reagent. However, unlike TREAT-HF and Olah′s Reagent, DMPU-HF is more tolerant of transition metals due to the reduced basicity of DMPU, compared to Triethylamine and Pyridine. Hammond and coworkers display its compatibility in transition-metal catalysis in gold-catalyzed mono- and difluoroination of alkynes.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 2 Dermal - Acute Tox. 2 Oral - Skin Corr. 1A

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shengzong Liang et al.
Journal of fluorine chemistry, 203, 136-139 (2018-02-13)
Bromofluorination reactions were performed by treating of a variety of unsaturated compounds with N-bromosuccinimide (NBS) and DMPU/HF as the fluorinating reagent. The DMPU/HF complex showed to be an efficient fluorinating reagent to convert alkenes into their corresponding bromofluoro compounds. It
Otome E Okoromoba et al.
Organic letters, 17(16), 3975-3977 (2015-08-12)
DMPU/HF (HF content 65 wt %/wt) is an ideal nucleophilic fluorination reagent for the diastereoselective synthesis of substituted 4-fluorotetrahydropyrans and 4-fluoropiperidines via a fluoro-Prins reaction. When compared to classical nucleophilic fluorination reagents like pyridine/HF, DMPU/HF gives both higher yields and
Otome E Okoromoba et al.
Journal of the American Chemical Society, 136(41), 14381-14384 (2014-09-27)
Hydrogen fluoride (HF) and selected nonbasic and weakly coordinating (toward cationic metal) hydrogen-bond acceptors (e.g., DMPU) can form stable complexes through hydrogen bonding. The DMPU/HF complex is a new nucleophilic fluorination reagent that has high acidity and is compatible with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service