Skip to Content
Merck
All Photos(1)

Key Documents

795291

Sigma-Aldrich

Phenofluor solution

0.1 M in toluene

Synonym(s):

1,3-Bis(2,6-diisopropylphenyl)-2,2-difluoro-4-imidazoline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H36F2N2
Molecular Weight:
426.58
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

form

liquid

Quality Level

concentration

0.1 M in toluene

density

0.865 g/mL at 25 °C

storage temp.

2-8°C

InChI

1S/C27H36F2N2/c1-17(2)21-11-9-12-22(18(3)4)25(21)30-15-16-31(27(30,28)29)26-23(19(5)6)13-10-14-24(26)20(7)8/h9-20H,1-8H3

InChI key

FQXXWTOSPDVNSG-UHFFFAOYSA-N

Application

Facile deoxyfluorination of phenols without preactivation (), along with highly chemoselective deoxyfluorination of highly functionalized late-stage intermediates ().

Legal Information

PhenoFluor is a trademark of SciFluor Life Sciences, LLC
PhenoFluor is a trademark of SciFluor Life Sciences, LLC

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

39.2 °F

Flash Point(C)

4 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Filippo Sladojevich et al.
Journal of the American Chemical Society, 135(7), 2470-2473 (2013-02-13)
An operationally simple protocol for the selective deoxyfluorination of structurally complex alcohols is presented. Several fluorinated derivatives of natural products and pharmaceuticals have been prepared to showcase the potential of the method for late-stage diversification and its functional group compatibility.
Pingping Tang et al.
Journal of the American Chemical Society, 133(30), 11482-11484 (2011-07-09)
An operationally simple ipso fluorination of phenols with a new deoxyfluorination reagent is presented.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service