Skip to Content
Merck
All Photos(1)

Key Documents

706930

Sigma-Aldrich

S-(11-Bromoundecyl) thioacetate

Synonym(s):

Thioacetic acid S-(11-bromoundecyl) ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H25BrOS
CAS Number:
Molecular Weight:
309.31
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

form

liquid

Quality Level

refractive index

n20/D 1.496

density

0.500 g/mL at 25 °C

SMILES string

CC(=O)SCCCCCCCCCCCBr

InChI

1S/C13H25BrOS/c1-13(15)16-12-10-8-6-4-2-3-5-7-9-11-14/h2-12H2,1H3

InChI key

JEDBIVNXPYJQIU-UHFFFAOYSA-N

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

>230.0 °F

Flash Point(C)

> 110 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oriol Penon et al.
Journal of colloid and interface science, 496, 100-110 (2017-02-20)
Photodynamic therapy (PDT) is a treatment of cancer by which tumour cells are destroyed using reactive oxygen species produced by photosensitizers following activation with visible or near infrared light. Successful PDT depends on the solubility and the targeting ability of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service