717398
(S)[(Sp)-2-(Diphenylphosphino)ferrocenyl]-4-isopropyloxazoline
97%
Synonym(s):
(Sp)-1-(Diphenylphosphino)-2-[(4S)-4-isopropyl-2-oxazolyl]ferrocene
About This Item
Recommended Products
Quality Level
Assay
97%
form
solid
optical activity
[α]20/D 366°, c = 0.1 in chloroform
mp
158-162 °C
functional group
ether
phosphine
SMILES string
[Fe].[CH]1[CH][CH][CH][CH]1.CC(C)[C@H]2COC(=N2)[C]3[CH][CH][CH][C]3P(c4ccccc4)c5ccccc5
InChI
1S/C23H23NOP.C5H5.Fe/c1-17(2)21-16-25-23(24-21)20-14-9-15-22(20)26(18-10-5-3-6-11-18)19-12-7-4-8-13-19;1-2-4-5-3-1;/h3-15,17,21H,16H2,1-2H3;1-5H;/t21-;;/m1../s1
InChI key
FELCHLOAEMNOQM-GHVWMZMZSA-N
Application
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service