Recommended Products
storage temp.
2-8°C
Quality Level
Application
Poly(dimethylsiloxane) (PDMS) is a commonly used material in microfluidic devices and other biomedical applications because of its high transparency, non-toxicity, and biocompatibility. However, the hydrophobic property of PDMS is a limiting factor for droplet-based microfluidics and several other applications. The use of this kit results in creating a durable hydrophilic PDMS surface. The resulting surface is wettable by hydrophilic inks and is appropriate for multiple printing applications. After treatment, water should completely wet the surface as the water contact angle is expected to be between 25 and 35°.
recommended
Product No.
Description
Pricing
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3
Supplementary Hazards
Storage Class Code
3 - Flammable liquids
Flash Point(F)
26.1 °F
Flash Point(C)
-3.3 °C
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Hydrophilic surface modification of PDMS microchannel for O/W and W/O/W emulsions
Micromachines, 6(10), 1445-1458 (2015)
Hydrophilic surface modification of PDMS for droplet microfluidics using a simple, quick, and robust method via PVA deposition
Microsystems & Nanoengineering, 24(3), 16091-16091 (2017)
Simple Surface Modification of Poly(dimethylsiloxane) via Surface Segregating Smart Polymers for Biomicrofluidics
Scientific reports, 9(1), 7377-7377 (2019)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service