All Photos(2)
About This Item
Empirical Formula (Hill Notation):
C11H10O3
CAS Number:
Molecular Weight:
190.20
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
97%
mp
65-69 °C (lit.)
functional group
ester
ketone
storage temp.
2-8°C
SMILES string
COC(=O)C1C(=O)Cc2ccccc12
InChI
1S/C11H10O3/c1-14-11(13)10-8-5-3-2-4-7(8)6-9(10)12/h2-5,10H,6H2,1H3
InChI key
RWSYHHRDYRDVQL-UHFFFAOYSA-N
Related Categories
Signal Word
Warning
Hazard Statements
Hazard Classifications
Acute Tox. 4 Oral
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service