Skip to Content
Merck
All Photos(1)

Key Documents

146293

Sigma-Aldrich

Ethyl (4-fluorobenzoyl)acetate

Synonym(s):

(p-Fluorobenzoyl)acetic acid ethyl ester, 4-Fluoro-ß-oxobenzenepropanoic acid ethyl ester, Ethyl 3-(4-fluorophenyl)-3-oxopropionate, Ethyl 4-fluorobenzoylacetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H4COCH2CO2C2H5
CAS Number:
Molecular Weight:
210.20
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

refractive index

n20/D 1.5040 (lit.)

Quality Level

bp

117-120 °C (lit.)

density

1.174 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)CC(=O)c1ccc(F)cc1

InChI

1S/C11H11FO3/c1-2-15-11(14)7-10(13)8-3-5-9(12)6-4-8/h3-6H,2,7H2,1H3

InChI key

SJUXLKYJKQBZLM-UHFFFAOYSA-N

General description

Ethyl (4-fluorobenzoyl)acetate on condensation with benzofurazan oxide yields 2-(carboethoxy)-3-(4′-fluoro)phenylquinoxaline1,4-dioxide.

Application

Reactant used as a precursor in:
  • Condensation reactions with diamines via C-C bond cleavage for synthesis of benzimidazoles and perimidines for possible use as antimalarial treatments
  • Base-promoted domino Michael addition / cyclization / elimination reactions for synthesis of hydroxybenzophenones
  • Oxidative cross-coupling with indoles via dioxygen activation
  • Cyclization of keto esters for synthesis of pyrones
  • Lewis base catalyzed hydrosilylation for synthesis of α-acetoxy β-amino acid derivatives
  • Conia-ene reactions for synthesis of methylenecyclopentane derivatives

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Comparative Use of Solvent-free KF-Al2O3 and K2CO3 in Acetone in the Synthesis of Quinoxaline 1, 4-Dioxide Derivatives Designed as Antimalarial Drug Candidates.
Lima LM, et al.
Journal of Heterocyclic Chemistry, 42(7), 1381-1385 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service