Skip to Content
Merck
All Photos(1)

Key Documents

387444

Sigma-Aldrich

Ethyl 3-nitrobenzoylacetate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H4COCH2CO2C2H5
CAS Number:
Molecular Weight:
237.21
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

powder

mp

75-77 °C (lit.)

SMILES string

CCOC(=O)CC(=O)c1cccc(c1)[N+]([O-])=O

InChI

1S/C11H11NO5/c1-2-17-11(14)7-10(13)8-4-3-5-9(6-8)12(15)16/h3-6H,2,7H2,1H3

InChI key

DSOJMGUVLXTQSE-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service