Skip to Content
Merck
All Photos(1)

Key Documents

601357

Sigma-Aldrich

Boron-10B trifluoride

95 atom % 10B

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
10BF3
Molecular Weight:
67.01
UNSPSC Code:
12142200

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

isotopic purity

95 atom % 10B

form

gas

mol wt

67.06 by atom % calculation

mass shift

M-1

SMILES string

F[10B](F)F

Packaging

All gas volumes are quoted at Standard Temperature and Pressure (STP) and not at Normal Temperature and Pressure (NTP). One STP Liter is 7.7% more gas than one NTP Liter.
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Eye Dam. 1 - Press. Gas Liquefied gas - Skin Corr. 1A - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Kidney, Respiratory system

Supplementary Hazards

Storage Class Code

2A - Gases

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service