Skip to Content
Merck
All Photos(1)

Key Documents

8.01663

Sigma-Aldrich

Boron trifluoride-methanol complex

(20% solution in methanol) for synthesis

Synonym(s):

BF3 methanol

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352301
NACRES:
NA.22
grade:
for synthesis
synthesis grade
vapor pressure:
4 hPa ( 20 °C)

grade

for synthesis
synthesis grade

Quality Level

vapor pressure

4 hPa ( 20 °C)

form

liquid

autoignition temp.

420 °C

expl. lim.

5.5 % (v/v) Methanol)

dilution

(for synthesis)

transition temp

flash point 16 °C

density

0.89 g/cm3 at 20 °C

storage temp.

2-30°C

General description

Boron trifluoride-methanol complex (BF-M) is Lewis acid boron trifluoride incoordination with methanol. BF-M is a powerful acidic catalyst for the esterification of fatty acids. In the deacetylation of acetanilides, the complex displayed excellent deprotection selectivity against amino-protecting groups.

Application

Boron trifluoride-methanol complex is used:
  • As a reagent for deprotection of N-acetyl derivatives (acetylated amines of cyanine-type dyes).
  • As a starting material in the automated radiosynthesis procedure for [18F]tetrafluoroborate.

Analysis Note

Assay BF3 (acidimetric): 14.0 - 16.0 %
Density (d 20 °C/ 4 °C): 0.880 - 0.895

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

60.8 °F

Flash Point(C)

16 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Boron trifluoride--methanol complex-mild and powerful reagent for deprotection of labile acetylated amines
Miltsov S, et al.
Tetrahedron Letters, 44(11), 2301-2303 (2003)
Comparison of transesterification methods for production of biodiesel from vegetable oils and fats
Demirbas A
Energy Conversion and Management , 49(1), 125-130 (2008)
Boron trifluoride--methanol complex. Mild and powerful reagent for deprotection of acetylated amines. Scope and selectivity
Miltsov S, et al.
Tetrahedron Letters, 57(6), 641-644 (2016)
High molar activity [18F] tetrafluoroborate synthesis for sodium iodide symporter imaging by PET
Soloviev D, et al.
EJNMMI radiopharmacy and chemistry, 7(1), 1-17 (2022)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service