Skip to Content
Merck
All Photos(1)

Key Documents

532320

Sigma-Aldrich

p-Toluenesulfonyl hydrazide, polymer-bound

100-200 mesh, extent of labeling: 2.0-3.0 mmol/g loading, 1 % cross-linked with divinylbenzene

Synonym(s):

Polystyrene sulfonyl hydrazide resin

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352125
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

crosslinking

1 % cross-linked with divinylbenzene

reaction suitability

reaction type: solution phase peptide synthesis

extent of labeling

2.0-3.0 mmol/g loading

particle size

100-200 mesh

functional group

hydrazine

SMILES string

[X]Cc1ccc(cc1)[S](=O)(=O)NN

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sharpiro, R.H.
Org. React., 23, 405-405 (1979)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service